missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMCR7L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543348
This item is not returnable.
View return policy
Description
Peptide sequence: GAAMLGIATL AVKRMYDRAI SAPTSPTRLS HSGKRSWEEP NWMGSPRLLN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Mitochondrial outer membrane protein which regulates mitochondrial fission. Promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface independently of the mitochondrial fission FIS1 and MFF proteins. Regulates DNM1L GTPase activity and DNM1L oligomerization. Binds ADP and can also bind GDP, although with lower affinity. Does not bind CDP, UDP, ATP, AMP or GTP. Inhibits DNM1L GTPase activity in the absence of bound ADP. Requires ADP to stimulate DNM1L GTPase activity and the assembly of DNM1L into long, oligomeric tubules with a spiral pattern, as opposed to the ring-like DNM1L oligomers observed in the absence of bound ADP. Does not require ADP for its function in recruiting DNM1L. [UniProt].
Specifications
| SMCR7L | |
| Polyclonal | |
| Unconjugated | |
| MIEF1 | |
| A230016E22; AI452372; dJ1104E15.3; HSU79252; MID51; Mief1; mitochondrial dynamic protein MID51; mitochondrial dynamic protein of 51 kDa; mitochondrial dynamic protein of 51 kDa homolog; mitochondrial dynamics protein MID51; Mitochondrial dynamics protein of 51 kDa; mitochondrial dynamics protein of 51 kDa homolog; Mitochondrial elongation factor 1; Smcr7l; SMCR7-like protein; smith-Magenis syndrome chromosomal region candidate gene 7 protein-like; Smith-Magenis syndrome chromosome region, candidate 7-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54471 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NQG6 | |
| MIEF1 | |
| Synthetic peptide directed towards the N-terminal of human SMCR7L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction