missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCP Antibody (OTI6G9), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74239AF405
This item is not returnable.
View return policy
Description
SMCP Monoclonal specifically detects SMCP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SMCP | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SMCP | |
| Full length recombinant protein of human SMCP (NP_109588) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI6G9 | |
| Western Blot, Immunohistochemistry | |
| HSMCSGEN1, MCSMGC26305, MCSPMGC26519, mitochondrial capsule selenoprotein, sperm mitochondria-associated cysteine-rich protein, sperm mitochondrial-associated cysteine-rich protein | |
| Mouse | |
| Affinity Purified | |
| Developmental Biology | |
| 4184.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction