missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCP Antibody (5C10D8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £435.00
Specifications
| Antigen | SMCP |
|---|---|
| Clone | 5C10D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669895
|
Novus Biologicals
NBP2-37253-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18182538
|
Novus Biologicals
NBP2-37253 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMCP Monoclonal specifically detects SMCP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SMCP | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| HSMCSGEN1, MCSMGC26305, MCSPMGC26519, mitochondrial capsule selenoprotein, sperm mitochondria-associated cysteine-rich protein, sperm mitochondrial-associated cysteine-rich protein | |
| SMCP | |
| IgG2b | |
| Affinity Purified |
| 5C10D8 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Developmental Biology | |
| PBS with 0.05% Sodium Azide | |
| 4184 | |
| Purified recombinant fragment of human SMCP (AA: FULL(1-116)) expressed in E. Coli. | |
| Primary | |
| 12.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title