missing translation for 'onlineSavingsMsg'
Learn More

SMCP Antibody (4F4), Novus Biologicals™

Product Code. 18322638 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
missing translation for 'unitSize'
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18322638 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 18322638 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' H00004184M04

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

Mouse Monoclonal Antibody

SMCP Monoclonal antibody specifically detects SMCP in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifica

Antigen SMCP
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4F4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14593
Gene Alias HSMCSGEN1, MCSMGC26305, MCSPMGC26519, mitochondrial capsule selenoprotein, sperm mitochondria-associated cysteine-rich protein, sperm mitochondrial-associated cysteine-rich protein
Host Species Mouse
Immunogen SMCP (AAH14593, 1 a.a. ~ 116 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MCDQTKHSKCCPAKGNQCCPPQQNQCCQSKGNQCCPPKQNQCCQPKGSQCCPPKHNHCCQPKPPCCIQARCCGLETKPEVSPLNMESEPNSPQTQDKGCQTQQQPHSPQNESRPSK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 4184
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.