missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCO4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25815-1-AP-150UL
This item is not returnable.
View return policy
Description
SMCO4 is a protein coding gene.Specifications
| SMCO4 | |
| Polyclonal | |
| Unconjugated | |
| SMCO4 | |
| C11orf75, FN5, Protein FN5, UPF0443 protein C11orf75 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56935 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NRQ5 | |
| SMCO4 | |
| SMCO4 Fusion Protein Ag22806 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction