missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMCHD1 Recombinant Rabbit Monoclonal Antibody (24GB2385)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557725
This item is not returnable.
View return policy
Description
SMCHD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
Specifications
| SMCHD1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SMCHD1 | |
| 4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23347, 316732 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB2385 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6NHR9 | |
| SMCHD1 | |
| A synthesized peptide derived from human SMCHD1 (aa 1800-2005). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction