missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMCHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577172
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Required for maintenance of X inactivation in females and hypermethylation of CpG islands associated with inactive X. Involved in a pathway that mediates the methylation of a subset of CpG islands slowly and requires the de novo methyltransferase DNMT3B. Required for DUX4 silencing in somatic cells.
Specifications
| SMCHD1 | |
| Polyclonal | |
| Unconjugated | |
| SMCHD1 | |
| 4931400A14Rik; AW554188; KIAA0650; mKIAA0650; MommeD1; RGD1307234; SMC hinge domain containing 1; SMC hinge domain-containing protein 1; Smchd1; structural maintenance of chromosomes flexible hinge domain containing 1; structural maintenance of chromosomes flexible hinge domain-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23347, 316732, 74355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NHR9, Q6P5D8 | |
| SMCHD1 | |
| Recombinant full length Human SMCHD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction