missing translation for 'onlineSavingsMsg'
Learn More

SMCC-DM1, MedChemExpress

Product Code. 16493248
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493248 10 mg 10mg
16492686 25 mg 25mg
16459796 50 mg 50mg
16455977 5 mg 5mg
16454516 100 mg 100mg
16400729 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16493248 Supplier Medchem Express Supplier No. HY101070
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SMCC-DM1 (DM1-SMCC) is a drug-linker conjugate composed of a potent microtubule-disrupting agent DM1 and a linker SMCC to make antibody drug conjugate (ADC).

SMCC-DM1 (DM1-SMCC) is a drug-linker conjugate composed of a potent microtubule-disrupting agent DM1 and a linker SMCC to make antibody drug conjugate (ADC).

Specifications

Chemical Name or Material SMCC-DM1
CAS 1228105-51-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C51H66ClN5O16S
Quantity 10 mg
Synonym DM1-SMCC
Solubility Information DMSO : 16.67 mg/mL (15.54 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(O[C@H]([C@@]1(C)[C@H]([C@@H]([C@@]2([H])OC(N[C@]3(O)C2)=O)C)O1)CC(N(C)C4=CC(C/C(C)=C/C=C/[C@H]3OC)=CC(OC)=C4Cl)=O)[C@@H](N(C(CCSC(C5=O)CC(N5CC6CCC(C(ON7C(CCC7=O)=O)=O)CC6)=O)=O)C)C
Molecular Weight (g/mol) 1072.61
Formula Weight 1072.61
Percent Purity 98.01%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.