missing translation for 'onlineSavingsMsg'
Learn More

SMC6L1 Antibody (2E6), Novus Biologicals™

Product Code. 18421159 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421159 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421159 Supplier Novus Biologicals Supplier No. H00079677M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMC6L1 Monoclonal antibody specifically detects SMC6L1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC6L1
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Chromatin Immunoprecipitation (ChIP)
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH39828
Gene Alias FLJ22116, hSMC6, SMC protein 6, SMC-6, SMC6 structural maintenance of chromosomes 6-like 1, SMC6 structural maintenance of chromosomes 6-like 1 (yeast), SMC6L1FLJ35534, structural maintenance of chromosomes 6, structural maintenance of chromosomes protein 6
Host Species Mouse
Immunogen SMC6L1 (AAH39828, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAKRKEENFSSPKNAKRPRQEELEDFDKDGDEDECKGTTLTAAEVGIIESIHLKNFMCHSMLGPFKFGSNVNFVVGNNGSGKSAVLTALIVGLGGRAVATNRGSSLKGFV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 79677
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.