missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC5 Polyclonal Antibody

SMC5 Polyclonal Antibody
Brand: Thermo Scientific PA5115931
This item is not returnable.
View return policy
Description
SMC5 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| SMC5 | |
| Polyclonal | |
| Unconjugated | |
| Smc5 | |
| hSMC5; KIAA0594; mKIAA0594; MLZ-453; mSMC5; Protein expressed in male leptotene and zygotene spermatocytes 453; SMC protein 5; Smc5; SMC-5; SMC5 structural maintenance of chromosomes 5-like 1; Smc5l1; structural maintenance of chromosomes 5; structural maintenance of chromosomes protein 5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 226026, 23137, 293967 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8CG46, Q8IY18 | |
| Smc5 | |
| A synthesized peptide derived from human SMC5(Accession Q8IY18), corresponding to amino acid residues E964-P1014. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction