missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA563037
This item is not returnable.
View return policy
Description
Immunogen sequence: EVPSKRKNSA PQLPLLQSSG PFVEGSIVRI SMENFLTYDI CEVSPGPHLN MIVGANGTGK SSIVCAICLG LAGKPAFMGR ADKVGFFVKR GCSRGMVEIE LFRASGNLVI TR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Core component of the SMC5-SMC6 complex, a complex involved in DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks.
Specifications
| SMC5 | |
| Polyclonal | |
| Unconjugated | |
| SMC5 | |
| SMC5L1, SMC protein 5, SMC-5, SMC5 structural maintece of chromosomes 5-like 1, hSMC5, structural maintece of chromosomes protein 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY18 | |
| SMC5 | |
| Recombit protein corresponding to Human SMC5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu