missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC4 Recombinant Rabbit Monoclonal Antibody (24GB2400)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557722
This item is not returnable.
View return policy
Description
SMC4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
Specifications
| SMC4 | |
| Recombinant Monoclonal | |
| 0.50 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CG47, Q9NTJ3 | |
| Smc4 | |
| A synthesized peptide derived from human SMC4 (aa 1200-1250). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2400 | |
| Unconjugated | |
| Smc4 | |
| 2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10051, 295107, 70099 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction