missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589539
This item is not returnable.
View return policy
Description
Immunogen sequence: MPRKGTQPST ARRREEGPPP PSPDGASSDA EPEPPSGRTE SPATAAAMTN EAGAPRLMIT HIVNQNFKSY AGEKILGPFH KRFSCIIGPN GSGKSNVIDS; Positive Samples: HT-29, MCF7, Raji, HeLa; Cellular Location: Chromosome, Cytoplasm, Nucleus.
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
Specifications
| SMC4 | |
| Polyclonal | |
| Unconjugated | |
| Smc4 | |
| 2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10051 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.15 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9NTJ3 | |
| Smc4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SMC4 (NP_0012756821). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction