missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35400-20ul
This item is not returnable.
View return policy
Description
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for ELISA,Western Blot
Specifications
| SMC4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SMC4 (NP_001275682.1).,, Sequence:, MPRKGTQPSTARRREEGPPPPSPDGASSDAEPEPPSGRTESPATAAAMTNEAGAPRLMITHIVNQNFKSYAGEKILGPFHKRFSCIIGPNGSGKSNVIDS | |
| 20 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 10051 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction