missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC3 Recombinant Rabbit Monoclonal Antibody (BLR099G)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544389
This item is not returnable.
View return policy
Description
SMC3 Recombinant Monoclonal Antibody for Western Blot, Flow, IP
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
Specifications
| SMC3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| BBS with 0.09% sodium azide; pH 7.8-8.6 | |
| Q9UQE7 | |
| SMC3 | |
| Peptide representing a region between residue 1175 and the C-terminus (residue 1217) of human Structural maintenance of chromosomes protein 3 (SMC3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| BLR099G | |
| Unconjugated | |
| SMC3 | |
| Bam; Bamacan; basement membrane chondroitin sulfate proteoglycan; basement membrane-associated chondroitin proteoglycan; BMH; CDLS3; chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); chromosome segregation protein SmcD; Chromosome-associated polypeptide; cspg6; HCAP; im:7142991; mad member-interacting protein 1; Mmip1; SMC protein 3; Smc3; SMC-3; SMC3 protein; SMC3L1; SmcD; structural maintenace of chromosomes 3; structural maintenance of chromosomes 3; structural maintenance of chromosomes protein 3; structural maintenance of chromosomes protein 3-like protein; wu:fb22e01; wu:fc30d07 | |
| Rabbit | |
| Purified | |
| RUO | |
| 9126 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction