missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC3 Recombinant Rabbit Monoclonal Antibody (24GB2155)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557721
This item is not returnable.
View return policy
Description
SMC3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
Specifications
| SMC3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SMC3 | |
| Bam; Bamacan; basement membrane chondroitin sulfate proteoglycan; basement membrane-associated chondroitin proteoglycan; BMH; CDLS3; chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); chromosome segregation protein SmcD; Chromosome-associated polypeptide; cspg6; HCAP; im:7142991; mad member-interacting protein 1; Mmip1; SMC protein 3; Smc3; SMC-3; SMC3 protein; SMC3L1; SmcD; structural maintenace of chromosomes 3; structural maintenance of chromosomes 3; structural maintenance of chromosomes protein 3; structural maintenance of chromosomes protein 3-like protein; wu:fb22e01; wu:fc30d07 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13006, 29486, 9126 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2155 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97690, Q9CW03, Q9UQE7 | |
| SMC3 | |
| A synthesized peptide derived from human SMC3 (aa 1150-1217). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction