missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19109
This item is not returnable.
View return policy
Description
SMC3 Polyclonal specifically detects SMC3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).
Specifications
| SMC3 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation (Negative) | |
| NP_005436.1 | |
| SMC3 | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human structural maintenance of chromosome 3 using the numbering given in entry NP_005436.1 (GeneID 9126). | |
| 0.1 mL | |
| Chromatin Research | |
| 9126 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction