missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | SMC3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug-4ug antibody for 400ug-600ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228169
|
Novus Biologicals
NBP3-35831-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230327
|
Novus Biologicals
NBP3-35831-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMC3 Polyclonal antibody specifically detects SMC3 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western BlotSpecifications
| SMC3 | |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Phospho Specific | |
| PBS (pH 7.3), 50% glycerol | |
| 9126 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug-4ug antibody for 400ug-600ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 909-1199 of human SMC3 (NP_005436.1).,, Sequence:, KEHMDAINHDTKELEKMTNRQGMLLKKKEECMKKIRELGSLPQEAFEKYQTLSLKQLFRKLEQCNTELKKYSHVNKKALDQFVNFSEQKEKLIKRQEELDRGYKSIMELMNVLELRKYEAIQLTFKQVSKNFSEVFQKLVPGGKATLVMKKGDVEGSQSQDEGEGSGESERGSGSQSSVPSVDQFTGVGIRVSFTGKQGEMREMQQLSGGQKSLVALALIFAIQKCDPAPFYLFDEIDQALDAQHRKAVSDMIMELAVHAQFITTTFRPELLESADKFYGVKFRNKVSHID | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title