missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35795-100ul
This item is not returnable.
View return policy
Description
SMC2 Polyclonal antibody specifically detects SMC2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| SMC2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CAP-E, CAPESMC2 structural maintenance of chromosomes 2-like 1 (yeast), Chromosome-associated protein E, FLJ10093, hCAP-EXCAP-E homolog, SMC protein 2, SMC-2, SMC2 (structural maintenance of chromosomes 2, yeast)-like 1, SMC2 structural maintenance of chromosomes 2-like 1, SMC2L1, structural maintenance of chromosomes (SMC) family member, structural maintenance of chromosomes 2, structural maintenance of chromosomes protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-900 of human SMC2 (NP_006435.2).,, Sequence:, EELAGLKNTAEKYRQLKQQWEMKTEEADLLQTKLQQSSYHKQQEELDALKKTIEESEETLKNTKEIQRKAEEKYEVLENKMKNAEAERERELKDAQKKLDCAKTKADASSKKMKEKQQEVEAITLELEELKREHTSYKQQLEAVNEAIKSYESQIEVMAAEVAKNKESVNKAQEEVTKQKEVITAQDTVIKAKYAEVAKHK | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 10592 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction