missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC1L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89966-25ul
This item is not returnable.
View return policy
Description
SMC1L2 Polyclonal specifically detects SMC1L2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SMC1L2 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q8NDV3 | |
| SMC1B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KSTMQILSVQAEENCLQTVNELMAKQQQLKDIRVTQNSSAEKVQTQIEEERKKFLAVDREVGKLQKEVVSIQTSLEQKRLEKHNLLLDCKVQDIEIILLSGSLDDIIEVEMGTEAESTQATIDIYEKEEAFEIDYSSL | |
| 25 μL | |
| Cell Cycle and Replication | |
| 27127 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bK268H5, bK268H5.5, FLJ43748, mitosis-specific chromosome segregation protein like protein beta, SMC protein 1B, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 2, SMC1 structural maintenance of chromosomes 1-like 2 (yeast), SMC-1B, SMC-1-beta, SMC1BETA, SMC1L2, structural maintenance of chromosomes 1B, structural maintenance of chromosomes protein 1B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction