missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588855
This item is not returnable.
View return policy
Description
Immunogen sequence: ILLKTAAPNL RALENLKTVR DKFQESTDAF EASRKEARLC RQEFEQVKKR RYDLFTQCFE HVSISIDQIY KKLCRNNSAQ VSSYIKEQTQ DQFQMIVISL KEEFYSRADA LIGIYPEYDD CMFSRVLTLD LSQYPDTEGQ ESSKRHGESR; Positive Samples: Rat testis; Cellular Location: Chromosome, Nucleus, centromere.
The SMC1B gene encodes a meiosis-specific subunit of the cohesin complex, which is essential for proper chromosome segregation during meiosis. It plays a critical role in sister chromatid cohesion and DNA recombination. The SMC1B protein, also known as SMC1 beta, forms a heterodimer with SMC3, which is necessary for the formation of the cohesin complex that holds sister chromatids together until anaphase. Studies have demonstrated that SMC1B is required for the activation of the spindle assembly checkpoint (SAC) during oocyte meiosis. The loss of SMC1B function leads to improper chromosome alignment and segregation, resulting in infertility due to meiotic arrest. Furthermore, the gene is expressed exclusively during meiosis, and mutations or deletions within SMC1B can cause sterility due to disruptions in meiotic progression.
Specifications
| SMC1B | |
| Polyclonal | |
| Unconjugated | |
| Smc1b | |
| Smc1b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1012-1161 of human SMC1B (NP_0012784301). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1.52 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| bK268H5; bK268H5.5; mitosis-specific chromosome segregation protein like protein beta; RP1-102D24.3; SMC; SMC protein 1B; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 structural maintenance of chromosomes 1-like 2; SMC1B; SMC-1B; SMC1BETA; SMC-1-beta; SMC1L2; structural maintenace of chromosomes 1B; structural maintenance of chromosomes 1B; structural maintenance of chromosomes protein 1B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 300121 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction