missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21695-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Sec7 domain family. The encoded protein is a guanine nucleotide exchange factor that regulates the recruitment of proteins to membranes by mediating GDP to GTP exchange. The encoded protein is localized to the Golgi apparatus and plays a role in vesicular trafficking by activating ADP ribosylation factor 1. The encoded protein has also been identified as an important host factor for viral replication. Multiple transcript variants have been observed for this gene.Specifications
| SMC1A | |
| Polyclonal | |
| Unconjugated | |
| SMC1A | |
| CDLS2, DXS423E, KIAA0178, SB1.8, SMC 1 alpha, SMC 1A, SMC protein 1A, SMC1, SMC1A, SMC1alpha, SMC1L1, SMCB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24061, 8243 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14683, Q9CU62 | |
| SMC1A | |
| SMC1A Fusion Protein Ag16286 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction