Learn More
SMC1A Mouse anti-Human, CoraLite 594, Clone: 1A5D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66987-100UL
Description
The SMC1A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMC1A. This antibody recognizes human, mouse, rat antigen. The SMC1A antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| SMC1A | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 8243 | |
| IgG2b |
| Immunofluorescence | |
| 1A5D2 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |