Learn More
Invitrogen™ SMC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117082
Description
Positive Samples: 22Rv1, NCI-H460, HeLa, 293T, SKOV3, Mouse liver, Mouse spleen, Mouse pancreas, Rat liver Immunogen sequence: MGFLKLIEIE NFKSYKGRQI IGPFQRFTAI IGPNGSGKSN LMDAISFVLG EKTSNLRVKT LRDLIHGAPV GKPAANRAFV SMVYSEEGAE DRTFARVIVG GSSEYKINNK VVQLHEYSEE LEKLGILIKA.
Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregatior defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
Specifications
| SMC1 | |
| Polyclonal | |
| Unconjugated | |
| SMC1A | |
| 14S cohesin SMC1 subunit; 5830426I24Rik; CDLS2; chromosome segregation protein SmcB; DKFZp686L19178; DXhXs423e; DXS423E; KIAA0178; MGC138332; mitosis-specific chromosome segregation protein like protein beta; mKIAA0178; RP6-29D12.1; Sb1.8; segregation of mitotic chromosomes 1; segregation of mitotic chromosomes b; segregation of mitotic chromosomes-like 1; SM1A; SMC; SMC (segregation of mitotic chromosomes 1)-like 1; SMC protein; SMC protein 1A; SMC protein 1B; SMC1; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 protein; SMC1 protein cohesin subunit; SMC1 structural maintenance of chromosomes 1-like 1; SMC1 structural maintenance of chromosomes 1-like 2; Smc1a; SMC-1A; smc1a.L; smc1aa; Smc1alpha; SMC-1-alpha; SMC1alpha protein; Smc1b; SMC-1B; SMC1BETA; SMC-1-beta; SMC1L1; Smc1l2; Smcb; SMC-like 1; SMC-protein; structural maintenace of chromosomes 1B; structural maintenance of chromosomes 1; structural maintenance of chromosomes 1 like 1; structural maintenance of chromosomes 1A; structural maintenance of chromosomes 1A a; structural maintenance of chromosomes 1A L homeolog; structural maintenance of chromosomes 1B; structural maintenance of chromosomes protein 1A; Structural maintenance of chromosomes protein 1B; Unknown (protein for IMAGE:8058685); XELAEV_18000030mg; xsmc1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 140557, 24061, 27127, 300121, 63996, 8243 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14683, Q8NDV3, Q920F6, Q9CU62, Q9Z1M9 | |
| SMC1A, SMC1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human SMC1 (NP_0062972). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.