missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC1, p Ser957 Antibody (SR1229), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | SMC1 |
|---|---|
| Clone | SR1229 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18687534
|
Novus Biologicals
NBP3-21544-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610954
|
Novus Biologicals
NBP3-21544-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMC1 Monoclonal antibody specifically detects SMC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| SMC1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A | |
| A synthesized peptide derived from human phospho-SMC1 [Ser957] (Uniprot #: Q14683) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1229 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 8243 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title