missing translation for 'onlineSavingsMsg'
Learn More

SMC1 Antibody (317705) [DyLight 350], Novus Biologicals™

Product Code. 30733337 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733337 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733337 Supplier Novus Biologicals Supplier No. FAB3294D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMC1 Monoclonal antibody specifically detects SMC1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC1
Applications Western Blot
Classification Monoclonal
Clone 317705
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A
Host Species Mouse
Immunogen E. coli-derived recombinant human SMC1, Met837-Gln1233, Accession # Q14683
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8243
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.