Learn More
LevGo™ SmartPans™ Cahn Style Aluminum Microbalance Accessory Pan
GREENER_CHOICE
Product Code. 30696096
Change view
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 100
Product Code. Sterility unitSize
30696096 Non-sterile Pack of 100
1 options
Product Code. 30696096 Supplier LevGo™ Supplier No. 41568100
Only null left
Add to basket
Add to basket
Accessory pans ideal for microbalances, organic micro elementary, and thermal analysis.
LevGo™ Cahn style accessory pans are compatible with most commercially available analyzers and microbalances and are ideal for organic, micro elementary, and thermal analysis, and for pharmaceutical applications.
  • Made of Ultra High Vacuum (UVH) Foil - A wettable and completely free from oils normally found on other grades of aluminum foil
  • Foil meets ASTM B-479, specification 3.1.4 and 10.3.1.
  • Compatible with most commercially available analyzers and microbalances
  • Foil and Pans are made in the USA
  • Dimensions Dia x H: 1 x 0.1 in. (25 x 3.6 mm)
  • Available in packs of 20 and 100
Specifications
Bottom Shape Round
Color Silver
Material UVH Foil
Sterility Non-sterile
Type Microbalance