missing translation for 'onlineSavingsMsg'
Learn More

LevGo™ Paper Weigh Boats

Product Code. 15893967
Click to view available options
Color:
Brown
White
Unit Size:
Pack of 250
Pack of 500
This item is not returnable. View return policy

Product Code. 15893967

Brand: LevGo™ 20203

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LevGo™ Paper Weigh Boats are a truly green solution to sample weighing. They are made from FSC™ certified (FSC™ mix credit) paper (Eco-IN) and when disposed of in the standard waste stream they will naturally biodegrade (Eco-OUT). Controls contents like a polystyrene weigh boat.

  • An alternative to regular weighing papers as they are much easier to handle
  • Made of grease proof paper that meets FDA 21 CFR 176.170 (components of paper in contact with aqueous and fatty foods) and with 2I CFR 176.180 (contact with dry foods)
  • Compatible with heat applications with a temperature limitation of 210°C

Specifications

Bottom Shape Flat
Color Brown
Disposable Single-use
Material Paper
Shape Square
Sterility Non-sterile
Type Weighing Boat
For Use With (Application) Meets FDA 21 CFR 176.170 (aqueous and fatty foods) and CFR 176.180 (contact with dry foods)
For Use With (Equipment) Scale
Size Large
Length (Metric) 95.25 mm
Width (Metric) 95.25 mm
Height (Metric) 25.4 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.