missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smart SolutionsTM v59 PharmaVetResiMix Kit 1, Dr. Ehrenstorfer
Click to view available options
Quantity:
4 x 1 mL
Unit Size:
4mL
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Albendazole∣Albendazole sulfone∣Albendazole sulfoxide∣Albendazole-2-aminosulfone∣Cinoxacin∣Ciprofloxacin∣Danofloxacin mesylate∣Dapsone∣Difloxacin hydrochloride∣Doramectin∣Emamectin benzoate∣Enoxacin∣Enrofloxacin∣Eprinomectin∣Febantel∣Fenbendazole∣Fleroxacin∣Flubendazole∣Flumequine∣Ivermectin∣Lomefloxacin hydrochloride∣Marbofloxacin∣Mebendazole∣Moxidectin∣N-Acetyl Sulfamethazine∣Nalidixic acid∣Netobimin Sodium Salt∣Norfloxacin∣Ofloxacin∣Oxfendazole∣Oxibendazole∣Oxolinic acid∣Pefloxacin methane sulfonate dihydrate∣Pipemidic acid∣Piromidic Acid∣Sarafloxacin hydrochloride∣Sulfabenzamide∣Sulfachloropyridazine∣Sulfadiazine∣Sulfadimethoxine∣Sulfadimidine∣Sulfadoxine∣Sulfaguanidine∣Sulfamerazine∣Sulfameter∣Sulfamethizole∣Sulfamethoxazole∣Sulfamethoxypyridazine∣Sulfamonomethoxine∣Sulfamoxol∣Sulfanilamide∣Sulfapyridine∣Sulfaquinoxaline∣Sulfathiazole∣Sulfisomidine∣Thiabendazole∣Thiabendazole-5-hydroxy∣Triclabendazole |
| CAS | 71751-41-2, 54965-21-8, 75184-71-3, 54029-12-8, 80983-34-2, 28657-80-9 |
| Analyte or Component Names | Abamectin∣Albendazole∣Albendazole sulfone∣Albendazole sulfoxide∣Albendazole-2-aminosulfone∣Cinoxacin∣Ciprofloxacin∣Danofloxacin mesylate∣Dapsone∣Difloxacin hydrochloride∣Doramectin∣Emamectin benzoate∣Enoxacin∣Enrofloxacin∣Eprinomectin∣Febantel∣Fenbendazole∣Fleroxacin∣Flubendazole∣Flumequine∣Ivermectin∣Lomefloxacin hydrochloride∣Marbofloxacin∣Mebendazole∣Moxidectin∣N-Acetyl Sulfamethazine∣Nalidixic acid∣Netobimin Sodium Salt∣Norfloxacin∣Ofloxacin∣Oxfendazole∣Oxibendazole∣Oxolinic acid∣Pefloxacin methane sulfonate dihydrate∣Pipemidic acid∣Piromidic Acid∣Sarafloxacin hydrochloride∣Sulfabenzamide∣Sulfachloropyridazine∣Sulfadiazine∣Sulfadimethoxine∣Sulfadimidine∣Sulfadoxine∣Sulfaguanidine∣Sulfamerazine∣Sulfameter∣Sulfamethizole∣Sulfamethoxazole∣Sulfamethoxypyridazine∣Sulfamonomethoxine∣Sulfamoxol∣Sulfanilamide∣Sulfapyridine∣Sulfaquinoxaline∣Sulfathiazole∣Sulfisomidine∣Thiabendazole∣Thiabendazole-5-hydroxy∣Triclabendazole |
| Shipping Condition | -18°C |
| Certifications/Compliance | ISO/IEC 17025 |
| Solution Type | Mixture |
| Solvent or Matrix | Methanol |
| Industry Type | Environmental Testing |
| Quantity | 4 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Dapsone 5 μg/mL∣Abamectin 10 μg/mL∣Albendazole 10 μg/mL∣Albendazole sulfone 10 μg/mL∣Albendazole sulfoxide 10 μg/mL∣Albendazole-2-aminosulfone 10 μg/mL∣Cinoxacin 10 μg/mL∣Ciprofloxacin 10 μg/mL∣Danofloxacin mesylate 10 μg/mL∣Difloxacin hydrochloride 10 μg/mL∣Doramectin 10 μg/mL∣Emamectin benzoate 10 μg/mL∣Enoxacin 10 μg/mL∣Enrofloxacin 10 μg/mL∣Eprinomectin 10 μg/mL∣Febantel 10 μg/mL∣Fenbendazole 10 μg/mL∣Fleroxacin 10 μg/mL∣Flubendazole 10 μg/mL∣Flumequine 10 μg/mL∣Ivermectin 10 μg/mL∣Lomefloxacin hydrochloride 10 μg/mL∣Marbofloxacin 10 μg/mL∣Mebendazole 10 μg/mL∣Moxidectin 10 μg/mL∣N-Acetyl Sulfamethazine 10 μg/mL∣Nalidixic acid 10 μg/mL∣Netobimin Sodium Salt 10 μg/mL∣Norfloxacin 10 μg/mL∣Ofloxacin 10 μg/mL∣Oxfendazole 10 μg/mL∣Oxibendazole 10 μg/mL∣Oxolinic acid 10 μg/mL∣Pefloxacin methane sulfonate dihydrate 10 μg/mL∣Pipemidic acid 10 μg/mL∣Piromidic Acid 10 μg/mL∣Sarafloxacin hydrochloride 10 μg/mL∣Sulfabenzamide 10 μg/mL∣Sulfachloropyridazine 10 μg/mL∣Sulfadiazine 10 μg/mL∣Sulfadimethoxine 10 μg/mL∣Sulfadimidine 10 μg/mL∣Sulfadoxine 10 μg/mL∣Sulfaguanidine 10 μg/mL∣Sulfamerazine 10 μg/mL∣Sulfameter 10 μg/mL∣Sulfamethizole 10 μg/mL∣Sulfamethoxazole 10 μg/mL∣Sulfamethoxypyridazine 10 μg/mL∣Sulfamonomethoxine 10 μg/mL∣Sulfamoxol 10 μg/mL∣Sulfanilamide 10 μg/mL∣Sulfapyridine 10 μg/mL∣Sulfaquinoxaline 10 μg/mL∣Sulfathiazole 10 μg/mL∣Sulfisomidine 10 μg/mL∣Thiabendazole 10 μg/mL∣Thiabendazole-5-hydroxy 10 μg/mL∣Triclabendazole 10 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction