Learn More
Moldex™ Smart FFP-Mask
Product Code. p-8013854
Click to view available options
For Use With (Application):
For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes
Nuisance Relief or Protection Against Ozone
Unit Size:
Pack of 10
Pack of 20
Product Code. 12352299
Brand: Moldex™ 248001
Only null left
Add to basket
Edge
Add to basket
Protects against Ozone, and nuisance relief
Moldex Smart Series FFP2 NR D non-valved dust mask.
Suited to most industrial applications this general purpose mask is strong and durable. The ActiveForm™ seal automatically adjusts to different face shapes and movements to give a secure seal.
Specifications
Certifications/Compliance EN 149:2001+A1:2009
Disposable Single-use
Product Type Smart FFP-Mask
For Use With (Application) For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes