missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Oligo Isolation Kit

Product Code. 30280704 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30280704 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280704 Supplier Thermo Scientific™ Supplier No. 60180101
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ SMART Digest™ Oligo Isolation Kits enable efficient digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids for accurate quantitation by LC-MS. This kit includes all necessary enzymes, buffers, and solid phase extraction (SPE) materials.

The SMART Digest™ Oligo Isolation Kit provides a streamlined solution for the digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids, ensuring precise quantitation by LC-MS. This kit includes all essential enzymes, buffers, and solid phase extraction (SPE) materials, offering a comprehensive and reliable choice for your laboratory needs.

  • Efficiently digests and extracts oligonucleotide-based drugs and metabolites from biofluids
  • Ensures precise quantitation by LC-MS
  • Includes all necessary enzymes for effective digestion
  • Comes with required buffers for optimal enzyme activity
  • Features solid phase extraction (SPE) components for high-throughput sample processing
  • Comprehensive kit designed to streamline your workflow
TRUSTED_SUSTAINABILITY

Specifications

Bed Weight 10 mg
Capture None
Cleavage Proteinase K cleaves aromatic, aliphatic, or hydrophobic residues in P1 and P1' positions at the carboxyl side of amino acid residues.
Enzyme Proteinase K
Format Non-Magnetic
For Use With (Application) Digestion and extraction of oligonucleotide-based drugs and metabolites from biofluids prior to quantitation by LC-MS
For Use With (Equipment) Universal
Particle Size 30 to 40 μm
Product Line SMART Digest
Sorbent Mixed Mode Weak Anion Exchanger, Polymeric
Type Oligo Isolation
Volume (Metric) 1 mL
Volume (Metric) Well 1 mL
Includes SMART Digest Soluble Proteinase K, Proteinase K Digest Buffer, 10 mg, 1 mL, 96-well, WAX SPE Plate, 25 mL SPE Elution Buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.