missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits

Product Code. 30276729 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Magnetic
Magnetic with SOLAμ SPE and collection plate
Magnetic, 10 Pk.
Non-magnetic
Non-magnetic with SOLAμ SPE and Collection Plate
Unit Size:
1 set
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
30276729 Magnetic, 10 Pk. 1 set
15648683 Non-magnetic Each
15658683 Non-magnetic with SOLAμ SPE and Collection Plate Each
15668683 Magnetic with SOLAμ SPE and collection plate Each
15678683 Magnetic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30276729 Supplier Thermo Scientific™ Supplier No. 60110104NW10PK
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain fast, simple, highly reproducible, and sensitive streptavidin immunocapture and trypsin protein digestion in a single well for your biomarker studies.

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits. These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The kit purifies various target biomolecules via the formation of strong streptavidin-biotin bond.

Solving the Challenges of Low-level Quantitation in Complex Matrices

The need to improve the ability to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging.

As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.

Immunocapture and Protein Digestion in a Single Well

These IA kits deliver a process that is:

  • Fast
  • Simple
  • Highly reproducible
  • Sensitive

Unique Design Reduces Workflow to Hours

In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.

TRUSTED_SUSTAINABILITY

Specifications

Description Magnetic, 10 Pk.
Capture Streptavidin
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Peptide Quantitation
Product Line SMART Digest
Type Immunocapture + Protein Digestion combined on a single bead
Includes Bulk Magnetic beads with immobilized streptavidin and trypsin for capture and digestion and SMART Digest buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.