Learn More
Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.
Brand:  Thermo Scientific™ 60111-103
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 15608693
  • £1390.79 / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits. These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The SMART Digest IA Protein A Kits target purification of immunoglobulins due to the strong affinity for binding to the Fc portion of the molecule.
Solving the challenges of low-level quantitation in complex matrices
The need to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging.
As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.
Immunocapture and Protein Digestion in a Single Well
These IA kits deliver a process that is:
  • Fast
  • Simple
  • Highly reproducible
  • Sensitive
Unique Design Reduces Workflow to Hours
In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.
Specifications
Specifications
SMART Digest IA Kit, Protein A magnetic with SOLAμ SPE and collection plate
C-terminal side of lysine and arginine amino acids
SMART Digest Resin, Magnetic
SMART Digest
Magnetic SMART Digest Resin, Collection Plate, SOLAμ SPE Plate
Protein A
Trypsin
Peptide Quantitation
Immunocapture + Protein Digestion combined on a single bead