missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SMARCD3 Monoclonal antibody specifically detects SMARCD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, ELISA
Specifications
Specifications
| Antigen | SMARCD3 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 5F12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_001003801 |
| Gene Alias | 60 kDa BRG-1/Brm-associated factor subunit C, 60kDa BRG-1/Brm associated factor subunit c, BAF60Cchromatin remodeling complex BAF60C subunit, BRG1-associated factor 60C, CRACD3, mammalian chromatin remodeling complex BRG1-associated factor 60C, MGC111010, Rsc6p, subfamily d, member 3, SWI/SNF complex 60 kDa subunit C, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 3, Swp73-like protein |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?