missing translation for 'onlineSavingsMsg'
Learn More

SMARCD3 Antibody (5B6), Novus Biologicals™

Product Code. 18336739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336739 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336739 Supplier Novus Biologicals Supplier No. H00006604M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMARCD3 Monoclonal antibody specifically detects SMARCD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5B6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001003801
Gene Alias 60 kDa BRG-1/Brm-associated factor subunit C, 60kDa BRG-1/Brm associated factor subunit c, BAF60Cchromatin remodeling complex BAF60C subunit, BRG1-associated factor 60C, CRACD3, mammalian chromatin remodeling complex BRG1-associated factor 60C, MGC111010, Rsc6p, subfamily d, member 3, SWI/SNF complex 60 kDa subunit C, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 3, Swp73-like protein
Host Species Mouse
Immunogen SMARCD3 (NP_001003801, 385 a.a. ~ 483 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGNPEEERRAEFYHQPWSQEAVSRYFYCKIQQRRQELEQSLVVRNT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6604
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.