missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566380
This item is not returnable.
View return policy
Description
Immunogen sequence: QYQRPGMSPGN RMPMAGLQVG PPAGSPFGAA APLRPGMPPT MMDPFRKRLL VPQAQPPMPA Q Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
SMARCD2 is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.
Specifications
| SMARCD2 | |
| Polyclonal | |
| Unconjugated | |
| SMARCD2 | |
| 60 kDa BRG-1/Brm-associated factor subunit B; AW322457; Baf60b; BRG1-associated factor 60B; chromatin remodeling complex BAF60B subunit; CRACD2; mammalian chromatin remodeling complex BRG1-associated factor 60B; PRO2451; Rsc6p; SMARCD2; SWI/SNF complex 60 kDa subunit B; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2; Swp73-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6603 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92925 | |
| SMARCD2 | |
| Recombinant Human SMARCD2. Recombinant protein control fragment (Product #RP-107055). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction