missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | SMARCD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657160
|
Novus Biologicals
NBP2-93825-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692651
|
Novus Biologicals
NBP2-93825-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCD1 Polyclonal antibody specifically detects SMARCD1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SMARCD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS (pH 7.3), 50% glycerol | |
| 6602 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 216-515 of human SMARCD1 (NP_003067.3). LEDSALSKYDATKQKRKFSSFFKSLVIELDKDLYGPDNHLVEWHRTATTQETDGFQVKRPGDVNVRCTVLLMLDYQPPQFKLDPRLARLLGIHTQTRPVIIQALWQYIKTHKLQDPHEREFVICDKYLQQIFESQRMKFSEIPQRLHALLMPPEPIIINHVISVDPNDQKKTACYDIDVEVDDTLKTQMNSFLLSTASQQEIATLDNKIHETIETINQLKTQREFMLSFARDPQGFINDWLQSQCRDLKTMTDVVGNPEEERRAEFYFQPWAQEAVCRYFYSKVQQRRQELEQALGIRNT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title