missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596015
This item is not returnable.
View return policy
Description
Immunogen sequence: PKLLGKLKDI IKRHQGTVTE DKNNASHVVY PVPGNLEEEE WVRPVMKRDK QVLLHWGYYP DSYDTWIPAS EIEASVEDAP TPEKPRKVHA KWILDTDTFN E; Positive Samples: U-87MG, Mouse spinal cord; Cellular Location: Nucleus.
Cdc73 is a tumor suppressor probably involved in transcriptional and post-transcriptional control pathways. Cdc73 is part of the PAF1 complex, a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9. PAF1 was first purified from yeast in association with RNA polymerase II and is believed to function in transcription elongation. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. In addition to coordinating events during transcription (initiation, promoter clearance, and elongation), hPAF also coordinates events in RNA quality control. Cdc73 may also be involved in cell cycle progression through the regulation of cyclin D1/PRAD1 expression.
Specifications
| SMARCC2 | |
| Polyclonal | |
| Unconjugated | |
| Smarcc2 | |
| 5930405J04Rik; BAF170; BRG1-associated factor 170; chromatin remodeling complex BAF170 subunit; CRACC2; DKFZp313D0632; LOC685179; mammalian chromatin remodeling complex BRG1-associated factor 170; Rsc8; similar to SWI/SNF-related matrix-associated actin-dependent regulator of chromatin c2; Smarcc2; SWI/SNF complex 170 kDa subunit; SWI/SNF complex subunit SMARCC2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 2; SWI3-like protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6601, 68094, 685179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PDG5, Q8TAQ2 | |
| Smarcc2 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SMARCC2 (NP_6207061). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction