missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCC2/BAF170 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93679-0.1ml
This item is not returnable.
View return policy
Description
SMARCC2/BAF170 Polyclonal antibody specifically detects SMARCC2/BAF170 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SMARCC2/BAF170 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000 | |
| BAF170DKFZp313D0632, BRG1-associated factor 170, chromatin remodeling complex BAF170 subunit, CRACC2, mammalian chromatin remodeling complex BRG1-associated factor 170, Rsc8, subfamily c, member 2, SWI/SNF complex 170 kDa subunit, SWI/SNF complex subunit SMARCC2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 2, SWI3-like protein | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SMARCC2 (NP_620706.1). PKLLGKLKDIIKRHQGTVTEDKNNASHVVYPVPGNLEEEEWVRPVMKRDKQVLLHWGYYPDSYDTWIPASEIEASVEDAPTPEKPRKVHAKWILDTDTFNE | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 6601 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction