missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCC1 Recombinant Rabbit Monoclonal Antibody (23GB4165), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | SMARCC1 |
|---|---|
| Clone | 23GB4165 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087713
![]() |
Invitrogen™
MA553700 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088040
|
Invitrogen™
MA553701 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCC1 Recombinant Monoclonal antibody specifically detects SMARCC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SMARCC1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P97496, Q92922 | |
| 20588, 301020, 6599 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4165 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI115498; BAF155; BRG1-associated factor 155; chromatin remodeling complex BAF155 subunit; CRACC1; mammalian chromatin remodeling complex BRG1-associated factor 155; Rsc8; Smarcc1; Srg3; SWI/SNF complex 155 kDa subunit; SWI/SNF complex subunit SMARCC1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily C member 1; SWI3; SWI3-related protein | |
| A synthesized peptide derived from SMARCC1/BAF155 (700-750AA). | |
| Primary | |
| -20°C | |
| SMARCC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title