missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SMARCB1/CEN22q FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 |
| Gene Alias | BAF47,INI1,RDT,SNF5,SNF5L1,Sfh1p,Snr1,hSNFS |
| Gene Symbol | SMARCB1 |
| Gene ID (Entrez) | 6598 |
| Product Type | DNA/RNA |
| Quantity | 200 μL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction