missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCAD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540914
This item is not returnable.
View return policy
Description
Peptide sequence: RANTPDSDIT EKTEDSSVPE TPDNERKASI SYFKNQRGIQ YIDLSSDSED Sequence homology: Cow: 92%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 86%; Rabbit: 92%; Rat: 92%.
SMARCAD1 belongs to the SNF2/RAD54 helicase family. It contains 2 CUE domains, 1 helicase ATP-binding domain, and 1 helicase C-terminal domain. It is a probable ATP-dependent DNA helicase.
Specifications
| SMARCAD1 | |
| Polyclonal | |
| Unconjugated | |
| SMARCAD1 | |
| ADERM; ATP-dependent helicase 1; ATP-dependent helicase SMARCAD1; AV081750; AW226546; D6Pas1; DKFZP762K2015; enhancer trap locus 1; enhancer trap locus homolog 1; Etl1; Etl-1; HEL1; hHEL1; KIAA1122; mKIAA1122; Smarcad1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A containing DEAD/H box 1; SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56916 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4L7 | |
| SMARCAD1 | |
| Synthetic peptide directed towards the N-terminal of human SMARCAD1 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction