missing translation for 'onlineSavingsMsg'
Learn More

SMARCAD1 Antibody, Novus Biologicals™

Product Code. 18349319 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349319 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349319 Supplier Novus Biologicals Supplier No. H00056916B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

SMARCAD1 Polyclonal antibody specifically detects SMARCAD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCAD1
Applications Western Blot, Immunocytochemistry, ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH45534.1
Gene Alias ATP-dependent helicase 1, DKFZP762K2015, EC 3.6.1, EC 3.6.4.12, HEL1, hHEL1, KIAA1122ETL1DKFZp762K2015, subfamily a, containing DEAD/H box 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A containing DEAD/H box 1, SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin
Host Species Mouse
Immunogen SMARCAD1 (AAH45534.1, 1 a.a. - 1028 a.a.) full-length human protein. MNLFNLDRFRFEKRNKIEEAPEATPQPSQPGPSSPISLSAEEENAEGEVSRANTPDSDITEKTEDSSVPETPDNERKASISYFKNQRGIQYIDLSSDSEDVVSPNCSNTVQEKTFNKDTVIIVSEPSEDEESQGLPTMARRNDDISELEDLSELEDLKDAKLQTLKELFPQRSDNDLLKLIESTSTMDGAIAAALLMFGDAGGGPRKRKLSSSSEPYEEDEFNDDQSIKKTRLDHGEESNESAESSNNWEKQESIVLKLQKEFPNFDKQELREVLKEHEWMYTEALESLKVFAEDQDMQYASQSEVPNGKEVSSRSQNYPKNATKTKLKQKFSMKAQNGFNKKRKKNVFNQKRVVEDSEYDSGSDVGSSLDEDYSSGEEVMEDGYKGKILHFLQDASIGELTLIPQCSQKKAQKITELRPFNSWEALFTKMSKTNGLSEDLIWHCKTLIQERDVVIRLMNKCEDISNKLTKQVTMLTGNGGGWNIEQPSILNQSLSLKPYQKVGLNWLALVHKHGLNGILADEMGLGKTIQAIAFLAYLYQEGNNGPHLIVVPASTIDNWLREVNLWCPTLKVLCYYGSQEERKQIRFNIHSRYEDYNVIVTTYNCAISSSDDRSLFRRLKLNYAIFDEGHMLKNMGSIRYQHLMTINANNRLLLTGTPVQNNLLELMSLLNFVMPHMFSSSTSEIRRMFSSKTKSADEQSIYEKERIAHAKQIIKPFILRRVKEEVLKQLPPKKDRIELCAMSEKQEQLYLGLFNRLKKSINNLVTEKNTEMCNVMMQLRKMANHPLLHRQYYTAEKLKEMSQLMLKEPTHCEANPDLIFEDMEVMTDFELHVLCKQYRHINNFQLDMDLILDSGKFRVLGCILSELKQKGDRVVLFSQFTMMLDILEVLLKHHQHRYLRLDGKTQISERIHLIDEFNTDMDIFVFLLSTKAGGLGINLTSANVVILHDIDCNPYNDKQAEDRCHRVGQTKEALVIKLISQGTIEESMLKINQQKLKLEQDMTTVDEGDEGSMPADIATLLKTSMGL
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 56916
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.