missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCA5/SNF2H Rabbit anti-Human, Mouse, Rat, Clone: S05-9D5, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | SMARCA5/SNF2H |
|---|---|
| Clone | S05-9D5 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μ/mL, Immunocytochemistry 2 μg/mL |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314455
|
Novus Biologicals
NBP3-15093-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309916
|
Novus Biologicals
NBP3-15093-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCA5/SNF2H Monoclonal antibody specifically detects SMARCA5/SNF2H in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunocytochemistrySpecifications
| SMARCA5/SNF2H | |
| Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μ/mL, Immunocytochemistry 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.6.1, EC 3.6.4.-, hISWI, hSNF2HSWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5, ISWI, SNF2Hsucrose nonfermenting-like 5, subfamily a, member 5, Sucrose nonfermenting protein 2 homolog, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 5, WCRF135 | |
| A synthetic peptide of human SMARCA5/SNF2H (Uniprot#: O60264) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S05-9D5 | |
| Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 8467 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title