missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCA5/SNF2H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55311
This item is not returnable.
View return policy
Description
SMARCA5/SNF2H Polyclonal specifically detects SMARCA5/SNF2H in Human samples. It is validated for Immunoprecipitation.
Specifications
| SMARCA5/SNF2H | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 3.6.1, EC 3.6.4.-, hISWI, hSNF2HSWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5, ISWI, SNF2Hsucrose nonfermenting-like 5, subfamily a, member 5, Sucrose nonfermenting protein 2 homolog, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 5, WCRF135 | |
| Rabbit | |
| 122 kDa | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate | |
| O60264 | |
| SMARCA5 | |
| The immunogen recognized by this antibody maps to a region between residue 1002 and the C-terminus (residue 1052) of human SNF2h/ISWI using the numbering given in entry NP_003592.2 (GeneID 8467). | |
| Affinity Purified | |
| RUO | |
| 8467 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction