missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SMARCA5 Monoclonal Antibody (14SNF-1G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA3055
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA3-055 detects SNF2h in human samples. MA3-055 has been succesfully used in western blot application. Immunogen is a synthetic peptide corresponding to residues 1028-1045 of human SNF2h.
SMARCA5 is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. SMARCA5 is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein encoded by this gene is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The encoded protein is similar in sequence to the Drosophila ISWI chromatin remodeling protein.
Specifikationer
| SMARCA5 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.05% sodium azide | |
| O60264 | |
| SMARCA5 | |
| Synthetic peptide coupled to ovalbumin corresponding to amino acids 1028-1045 of human SNF2h. | |
| RUO | |
| 8467 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 14SNF-1G2 | |
| Unconjugated | |
| SMARCA5 | |
| 4933427E24Rik; D030040M08Rik; D330027N15Rik; hISWI; hSNF2H; ISWI; MommeD4; mSnf2h; SMARCA5; SNF2H; sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5; WCRF135 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering