missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMARCA2 Recombinant Rabbit Monoclonal Antibody (K01_2P50)

Product Code. 30830363
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830363 50 μL
30828561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830363 Supplier Invitrogen™ Supplier No. MA565667
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: STDFPQEGMH QMHKPI.

SMARCA2 encodes a protein that is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2P50
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene SMARCA2
Gene Accession No. P51531
Gene Alias 2610209L14Rik; ATP-dependent helicase SMARCA2; BAF190; Baf190b; brahma; BRG1-associated factor 190B; BRM; FLJ36757; global transcription activator homologous sequence; hBRM; hSNF2a; MGC74511; NCBRS; Probable global transcription activator SNF2L2; protein brahma homolog; SMARCA2; SNF2; SNF2/SWI2-like protein 2; SNF2A; SNF2alpha; SNF2-alpha; Snf2l2; SNF2LA; Sth1p; subunit of the SWI/SNF chromatin remodeling complex; sucrose nonfermenting 2-like protein 2; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 2; SWI2
Gene Symbols SMARCA2
Host Species Rabbit
Immunogen A synthetic peptide of human SMARCA2/BRM (aa 62-77).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6595
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.