missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCA1 Antibody (2H7B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61879
This item is not returnable.
View return policy
Description
SMARCA1 Monoclonal specifically detects SMARCA1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SMARCA1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| ATP-dependent helicase SMARCA1, DKFZp686D1623, EC 3.6.1, EC 3.6.4.-, FLJ41547, global transcription activator homologous sequence, Nucleosome-remodeling factor subunit SNF2L, NURF140, probable global transcription activator SNF2L1, SNF2L, SNF2L1ISWI, SNF2LB, SNF2-like 1, SNF2LT, subfamily a, member 1, sucrose nonfermenting 2-like protein 1, SWI, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 1, SWI2 | |
| Mouse | |
| 122.6 kDa | |
| 0.1 mL | |
| 6594 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2H7B8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SMARCA1 | |
| Purified recombinant fragment of human SMARCA1 (AA: 933-1070) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction