missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMAP2 Monoclonal Antibody (OTI9B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527417
This item is not returnable.
View return policy
Description
SMAP2 Monoclonal Antibody for Western Blot, IHC (P)
SMAP2 (Small ArfGAP2) is a Protein Coding gene. Among its related pathways are Endocytosis. Gene Ontology (GO) annotations related to this gene include GTPase activator activity. An important paralog of this gene is SMAP1.
Specifications
| SMAP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WU79 | |
| Smap2 | |
| Full length human recombinant protein of SMAP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9B6 | |
| Unconjugated | |
| Smap2 | |
| 1810031K02Rik; AA408614; RGD1308418; small ArfGAP 2; small ArfGAP2; SMAP1L; SMAP2; stromal membrane-associated GTPase-activating protein 2; Stromal membrane-associated protein 1-like; Stromal membrane-associated protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64744 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction