missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Small Volume™ Advanced TC™ Treated 384 μClear Well Cell Culture Microplates
Description
- Non-pyrogenic, non-cytotoxic
- Improved cell adherence
- Consistent cell attachment
- Homogenous cell growth
- In-vivo like morphology
- Increased cell yield
- Permits usage of serum-reduced or serum free media<
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Content And Storage | Room Temperature |
| Culture Area | 2 to 7 mm2 |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Packaging | 15/Pk., 60/Cs. |
| Color | Black, Clear Bottom |
| Product Type | Microplate |
| Material | Polystyrene |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?